Product Details

SNP ID
rs926385
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:209585387 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATTACAGCATCATAAGGATGAAG[A/G]TGGAGATGAGTGACATCCCAGCAAC
Phenotype
MIM: 614994
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAMK1G PubMed Links

Gene Details

Gene
CAMK1G
Gene Name
calcium/calmodulin dependent protein kinase IG
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020439.2 Intron NP_065172.1
XM_017001866.1 Intron XP_016857355.1
XM_017001867.1 Intron XP_016857356.1

View Full Product Details