Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001320046.1 | 2406 | Intron | NP_001306975.1 | ||
NM_032630.2 | 2406 | Intron | NP_116019.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001267827.1 | 2406 | Missense Mutation | TTA,TTC | L731F | NP_001254756.1 |
NM_001267828.1 | 2406 | Missense Mutation | TTA,TTC | L731F | NP_001254757.1 |
NM_018335.4 | 2406 | Missense Mutation | TTA,TTC | L847F | NP_060805.3 |
XM_006720203.3 | 2406 | Missense Mutation | TTA,TTC | L759F | XP_006720266.2 |
XM_011536945.1 | 2406 | Missense Mutation | TTA,TTC | L881F | XP_011535247.1 |
XM_011536946.2 | 2406 | Missense Mutation | TTA,TTC | L831F | XP_011535248.1 |
XM_011536948.2 | 2406 | Missense Mutation | TTA,TTC | L731F | XP_011535250.1 |
XM_011536949.2 | 2406 | Intron | XP_011535251.1 | ||
XM_011536950.2 | 2406 | Intron | XP_011535252.1 | ||
XM_017021447.1 | 2406 | Missense Mutation | TTA,TTC | L797F | XP_016876936.1 |
XM_017021448.1 | 2406 | Intron | XP_016876937.1 | ||
XM_017021449.1 | 2406 | Missense Mutation | TTA,TTC | L512F | XP_016876938.1 |
XM_017021450.1 | 2406 | Intron | XP_016876939.1 | ||
XM_017021451.1 | 2406 | Intron | XP_016876940.1 |