Product Details

SNP ID
rs1558644
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:102074850 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGGTGATTTTTATTTTCTTCTTT[A/G]CTAAACATTTCTGCATTTCATGAAG
Phenotype
MIM: 147810
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL1R1 PubMed Links

Gene Details

Gene
IL1R1
Gene Name
interleukin 1 receptor type 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000877.3 Intron NP_000868.1
NM_001288706.1 Intron NP_001275635.1
NM_001320978.1 Intron NP_001307907.1
NM_001320980.1 Intron NP_001307909.1
NM_001320981.1 Intron NP_001307910.1
NM_001320982.1 Intron NP_001307911.1
NM_001320983.1 Intron NP_001307912.1
NM_001320984.1 Intron NP_001307913.1
NM_001320985.1 Intron NP_001307914.1
NM_001320986.1 Intron NP_001307915.1
XM_005263930.4 Intron XP_005263987.1
XM_005263934.4 Intron XP_005263991.1
XM_011511115.2 Intron XP_011509417.1
XM_011511116.1 Intron XP_011509418.1
XM_011511117.1 Intron XP_011509419.1
XM_011511118.2 Intron XP_011509420.1
XM_017003989.1 Intron XP_016859478.1

View Full Product Details