Product Details

SNP ID
rs1056709
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:52498046 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTTTTCCTCTTCAACACATTTTT[A/T]AAAAATAGACCTCTAAGATGATGCT
Phenotype
MIM: 608815 MIM: 608485
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
EFHC1 PubMed Links

Gene Details

Gene
EFHC1
Gene Name
EF-hand domain containing 1
There are no transcripts associated with this gene.

Gene
TRAM2
Gene Name
translocation associated membrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012288.3 6288 UTR 3 NP_036420.1
XM_011515005.2 6288 UTR 3 XP_011513307.1

View Full Product Details