Product Details
- SNP ID
-
rs1796320
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:87783945 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGCACAGCTGCTTCTTTGTTTCCAC[A/G]GCAGTTTGCTCTTTCTCTGATTCTT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MKRN9P
PubMed Links
Gene Details
- Gene
- MKRN9P
- Gene Name
- makorin ring finger protein 9, pseudogene
There are no transcripts associated with this gene.
View Full Product Details