Product Details
- SNP ID
-
rs1215499
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
23
- Location
-
Chr.1:89632730 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTGTGGTTTTCCGCGCATTGTGAGG[A/G]ATGAGGGGTGGAGGTGGTATTAGAC
- Phenotype
-
MIM: 612889
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FLJ27354
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113690266] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FLJ27354
- Gene Name
- uncharacterized LOC400761
There are no transcripts associated with this gene.
- Gene
- LRRC8C
- Gene Name
- leucine rich repeat containing 8 family member C
View Full Product Details