Product Details

SNP ID
rs1045813
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:21460175 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAGGAAGAAAAAAACTCAATGAA[A/G]TTCCGAGGCAGAGGATCTATCAACC
Phenotype
MIM: 607466
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB2B PubMed Links

Gene Details

Gene
RAB2B
Gene Name
RAB2B, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163380.1 1700 UTR 3 NP_001156852.1
NM_032846.3 1700 UTR 3 NP_116235.2
XM_017021710.1 1700 UTR 3 XP_016877199.1
XM_017021711.1 1700 UTR 3 XP_016877200.1
XM_017021712.1 1700 UTR 3 XP_016877201.1

View Full Product Details