Product Details

SNP ID
hCV8921343
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6570131 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAATTTTCATCAAGAAATTTGCTG[G/T]GGTTTTTTATGCTTTTGGTTTTTTC
Phenotype
MIM: 603277 MIM: 164031
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CHD4 PubMed Links

Gene Details

Gene
CHD4
Gene Name
chromodomain helicase DNA binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001273.3 6341 UTR 3 NP_001264.2
NM_001297553.1 6341 UTR 3 NP_001284482.1
Gene
NOP2
Gene Name
NOP2 nucleolar protein
There are no transcripts associated with this gene.

View Full Product Details