Product Details
- SNP ID
-
rs1049562
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:20472356 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACACTGCAGAATGGCTTCTGTCTCA[C/T]ACTAAGCACCGACCTCAAGTTGCAA
- Phenotype
-
MIM: 164050
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PNP
PubMed Links
Gene Details
- Gene
- PNP
- Gene Name
- purine nucleoside phosphorylase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000270.3 |
206 |
Silent Mutation |
CAC,CAT |
H20H |
NP_000261.2 |
View Full Product Details