Product Details

SNP ID
rs927493
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24147574 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGACCGTGGTCTGAGTGACCTGGG[A/G]CGGCTGCGTGGGCCGGGGTGGGCCT
Phenotype
MIM: 600654 MIM: 602161 MIM: 612487
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
EMC9 PubMed Links
Additional Information
For this assay, SNP(s) [rs115578731] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
EMC9
Gene Name
ER membrane protein complex subunit 9
There are no transcripts associated with this gene.

Gene
MIR7703
Gene Name
microRNA 7703
There are no transcripts associated with this gene.

Gene
PSME1
Gene Name
proteasome activator subunit 1
There are no transcripts associated with this gene.

Gene
PSME2
Gene Name
proteasome activator subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002818.2 125 Intron NP_002809.2
XM_006720213.1 125 Intron XP_006720276.1
Gene
RNF31
Gene Name
ring finger protein 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001310332.1 125 Intron NP_001297261.1
NM_017999.4 125 UTR 5 NP_060469.4

View Full Product Details