Product Details
- SNP ID
-
rs854191
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:45393217 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCACATGTTATATATTTTTGTTTCT[C/T]ATCTGCCTCCCTTATTGTTTGTTTC
- Phenotype
-
MIM: 604544
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LARS2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs149219761] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LARS2
- Gene Name
- leucyl-tRNA synthetase 2, mitochondrial
View Full Product Details