Product Details

SNP ID
rs862701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57352569 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGGAATGGCTGGCCGAGGTGTCT[C/T]GAGTGTGGTAAAAGTCGTGGGAGAT
Phenotype
MIM: 613840
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF304 PubMed Links

Gene Details

Gene
ZNF304
Gene Name
zinc finger protein 304
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290318.1 Intron NP_001277247.1
NM_001290319.1 Intron NP_001277248.1
NM_020657.3 Intron NP_065708.2
XM_011527145.2 Intron XP_011525447.1
XM_017027024.1 Intron XP_016882513.1

View Full Product Details