Product Details
- SNP ID
-
rs1265084
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:31143012 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTGCTGCTGCTACAGGTGCAGATGC[C/T]GAACACTCTGGAGGCCTGGGGCTGG
- Phenotype
-
MIM: 605310
MIM: 613525
MIM: 613526
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CCHCR1
PubMed Links
Gene Details
- Gene
- CCHCR1
- Gene Name
- coiled-coil alpha-helical rod protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001105563.1 |
2378 |
Silent Mutation |
TCA,TCG |
S778S |
NP_001099033.1 |
NM_001105564.1 |
2378 |
Silent Mutation |
TCA,TCG |
S814S |
NP_001099034.1 |
NM_019052.3 |
2378 |
Silent Mutation |
TCA,TCG |
S725S |
NP_061925.2 |
XM_011514702.2 |
2378 |
Silent Mutation |
TCA,TCG |
S787S |
XP_011513004.1 |
XM_011514703.1 |
2378 |
Silent Mutation |
TCA,TCG |
S725S |
XP_011513005.1 |
XM_011514704.2 |
2378 |
Silent Mutation |
TCA,TCG |
S689S |
XP_011513006.1 |
XM_011514705.1 |
2378 |
Silent Mutation |
TCA,TCG |
S689S |
XP_011513007.1 |
XM_017010961.1 |
2378 |
Silent Mutation |
TCA,TCG |
S823S |
XP_016866450.1 |
XM_017010962.1 |
2378 |
Silent Mutation |
TCA,TCG |
S725S |
XP_016866451.1 |
XM_017010963.1 |
2378 |
Silent Mutation |
TCA,TCG |
S725S |
XP_016866452.1 |
XM_017010964.1 |
2378 |
Silent Mutation |
TCA,TCG |
S725S |
XP_016866453.1 |
XM_017010965.1 |
2378 |
Silent Mutation |
TCA,TCG |
S725S |
XP_016866454.1 |
XM_017010966.1 |
2378 |
Silent Mutation |
TCA,TCG |
S725S |
XP_016866455.1 |
XM_017010967.1 |
2378 |
Silent Mutation |
TCA,TCG |
S725S |
XP_016866456.1 |
XM_017010968.1 |
2378 |
Silent Mutation |
TCA,TCG |
S725S |
XP_016866457.1 |
XM_017010969.1 |
2378 |
Silent Mutation |
TCA,TCG |
S689S |
XP_016866458.1 |
XM_017010970.1 |
2378 |
Silent Mutation |
TCA,TCG |
S449S |
XP_016866459.1 |
- Gene
- PSORS1C1
- Gene Name
- psoriasis susceptibility 1 candidate 1
There are no transcripts associated with this gene.
- Gene
- PSORS1C2
- Gene Name
- psoriasis susceptibility 1 candidate 2
There are no transcripts associated with this gene.
View Full Product Details