Product Details

SNP ID
rs1043292
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:144159578 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GATAATTTACAGTAATGTCAAATTT[C/T]TGGAGTGCAAACTCATACTCTACAT
Phenotype
MIM: 611483
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
YIPF5 PubMed Links

Gene Details

Gene
YIPF5
Gene Name
Yip1 domain family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024947.3 1783 UTR 3 NP_001020118.1
NM_001271732.1 1783 UTR 3 NP_001258661.1
NM_030799.8 1783 UTR 3 NP_110426.4

View Full Product Details