Product Details
- SNP ID
-
rs4658297
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
26
- Location
-
Chr.1:92250017 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- CCCCCAACTTTTTTTTTTTCTTTTT[A/G]CTTTGCATTCACCTCTGGGTTTATA
- Phenotype
-
MIM: 601749
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C1orf146
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs548465107] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C1orf146
- Gene Name
- chromosome 1 open reading frame 146
There are no transcripts associated with this gene.
- Gene
- GLMN
- Gene Name
- glomulin, FKBP associated protein
View Full Product Details