Product Details

SNP ID
rs1055543
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:19510959 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGAGTTACACGATATTTATTTGCA[C/T]ACAGCTTCGTGATGTACAATGTGTG
Phenotype
MIM: 607026
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NAV2 PubMed Links

Gene Details

Gene
NAV2
Gene Name
neuron navigator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111018.1 Intron NP_001104488.1
NM_001111019.2 Intron NP_001104489.1
NM_001244963.1 Intron NP_001231892.1
NM_145117.4 Intron NP_660093.2
NM_182964.5 Intron NP_892009.3
XM_005253214.4 Intron XP_005253271.1
XM_006718364.3 Intron XP_006718427.1
XM_006718365.3 Intron XP_006718428.1
XM_006718366.3 Intron XP_006718429.1
XM_006718367.3 Intron XP_006718430.1
XM_006718368.3 Intron XP_006718431.1
XM_006718369.2 Intron XP_006718432.1
XM_011520444.2 Intron XP_011518746.1
XM_011520445.2 Intron XP_011518747.1
XM_011520446.2 Intron XP_011518748.1
XM_011520447.2 Intron XP_011518749.1
XM_011520448.2 Intron XP_011518750.1
XM_011520449.2 Intron XP_011518751.1
XM_011520450.2 Intron XP_011518752.1
XM_011520451.2 Intron XP_011518753.1
XM_011520452.2 Intron XP_011518754.1
XM_011520453.2 Intron XP_011518755.1
XM_011520454.2 Intron XP_011518756.1
XM_017018520.1 Intron XP_016874009.1
XM_017018521.1 Intron XP_016874010.1
XM_017018522.1 Intron XP_016874011.1
XM_017018523.1 Intron XP_016874012.1
XM_017018524.1 Intron XP_016874013.1
XM_017018525.1 Intron XP_016874014.1
XM_017018526.1 Intron XP_016874015.1
Gene
NAV2-AS4
Gene Name
NAV2 antisense RNA 4
There are no transcripts associated with this gene.

Gene
NAV2-AS5
Gene Name
NAV2 antisense RNA 5
There are no transcripts associated with this gene.

View Full Product Details