Product Details

SNP ID
rs1053513
Assay Type
Functionally tested
NCBI dbSNP Submissions
37
Location
Chr.1:229516840 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTTTACTAATTCAAAGAACACTA[C/T]ATTCATTCATATCAATTTTATTCAT
Phenotype
MIM: 605454 MIM: 607613
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABCB10 PubMed Links

Gene Details

Gene
ABCB10
Gene Name
ATP binding cassette subfamily B member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012089.2 4370 Intron NP_036221.2
XM_011544135.2 4370 UTR 3 XP_011542437.1
XM_011544136.2 4370 Intron XP_011542438.1
Gene
LOC101927478
Gene Name
uncharacterized LOC101927478
There are no transcripts associated with this gene.

Gene
NUP133
Gene Name
nucleoporin 133
There are no transcripts associated with this gene.

View Full Product Details