Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001190266.1 | Intron | NP_001177195.1 | |||
NM_001190267.1 | Intron | NP_001177196.1 | |||
NM_017974.3 | Intron | NP_060444.3 | |||
NM_030803.6 | Intron | NP_110430.5 | |||
NM_198890.2 | Intron | NP_942593.2 | |||
XM_005246082.2 | Intron | XP_005246139.1 | |||
XM_005246084.2 | Intron | XP_005246141.1 | |||
XM_005246086.2 | Intron | XP_005246143.1 | |||
XM_006712608.2 | Intron | XP_006712671.1 |