Product Details
- SNP ID
-
rs1486865
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:112544495 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TAACGAAAAAGGGAAAGTGAACTTG[A/G]CAGTTTTTAGTAGAATTGAAGGCTC
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZGRF1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116804237] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZGRF1
- Gene Name
- zinc finger GRF-type containing 1
View Full Product Details