Product Details

SNP ID
rs11600628
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:64208555 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TCAGCCTGGCCGCCCCGGGCCAGTC[T/C]CTGGCCTCCTGGAGCAGCAGCGGGT
Phenotype
MIM: 607901 MIM: 605063
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
FERMT3 PubMed Links

Gene Details

Gene
FERMT3
Gene Name
fermitin family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031471.5 Intron NP_113659.3
NM_178443.2 Intron NP_848537.1
XM_011545294.2 Intron XP_011543596.1
XM_011545295.2 Intron XP_011543597.1
XM_017018398.1 Intron XP_016873887.1
XM_017018399.1 Intron XP_016873888.1
Gene
STIP1
Gene Name
stress induced phosphoprotein 1
There are no transcripts associated with this gene.

View Full Product Details