Product Details

SNP ID
rs2278478
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:232526753 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TACCAGGATAGCCATGGAGGAAGCT[A/G]GAAGCCCCCACCTGGCCTATGGCCA
Phenotype
MIM: 100720 MIM: 613858
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHRND PubMed Links

Gene Details

Gene
CHRND
Gene Name
cholinergic receptor nicotinic delta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000751.2 Intron NP_000742.1
NM_001256657.1 Intron NP_001243586.1
NM_001311195.1 Intron NP_001298124.1
NM_001311196.1 Intron NP_001298125.1
XM_011510524.2 Intron XP_011508826.1
Gene
PRSS56
Gene Name
protease, serine 56
There are no transcripts associated with this gene.

View Full Product Details