Product Details

SNP ID
rs4496923
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:56020308 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATCAAGAAAGAGACCAAAGGTTAA[A/G]CATTCTTCCATGAAAGGCAGTTGAA
Phenotype
MIM: 172480
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PSPH PubMed Links

Gene Details

Gene
PSPH
Gene Name
phosphoserine phosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004577.3 Intron NP_004568.2
XM_005271773.1 Intron XP_005271830.1
XM_005271774.1 Intron XP_005271831.1
XM_005271775.1 Intron XP_005271832.1
XM_005271776.1 Intron XP_005271833.1
XM_006715760.1 Intron XP_006715823.1
XM_006715761.1 Intron XP_006715824.1
XM_011515461.1 Intron XP_011513763.1
XM_017012466.1 Intron XP_016867955.1
XM_017012467.1 Intron XP_016867956.1
XM_017012468.1 Intron XP_016867957.1
XM_017012469.1 Intron XP_016867958.1

View Full Product Details