Product Details

SNP ID
rs11629718
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:92471665 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGCTCAGCTGGACATTGGAACCAC[G/T]GGAGGAGCTTTAAAACCTCCTGAAA
Phenotype
MIM: 602546
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C15orf32 PubMed Links

Gene Details

Gene
C15orf32
Gene Name
chromosome 15 open reading frame 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301106.1 5904 Intron NP_001288035.1
NM_153040.2 5904 Intron NP_694585.1
XM_011521257.2 5904 UTR 5 XP_011519559.1
Gene
LOC104613533
Gene Name
uncharacterized LOC104613533
There are no transcripts associated with this gene.

Gene
ST8SIA2
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 2
There are no transcripts associated with this gene.

View Full Product Details