Product Details

SNP ID
rs1454503
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:3013503 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTGTAGAGAAGTACTGTTGAAATGG[A/G]AAACTTACAGCATTAAGAGTTGTCA
Phenotype
MIM: 300586
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARSH PubMed Links

Gene Details

Gene
ARSH
Gene Name
arylsulfatase family member H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011719.1 Intron NP_001011719.1

View Full Product Details