Product Details
- SNP ID
-
rs1042164
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:13153584 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGCAGCCTGAGCGACGGCGGGGACG[C/T]TGGACTGGTCCCGAGCAAGAAAGCC
- Phenotype
-
MIM: 603765
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IER2
PubMed Links
Gene Details
- Gene
- IER2
- Gene Name
- immediate early response 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004907.2 |
726 |
Missense Mutation |
GCT,GTT |
A133V |
NP_004898.2 |
- Gene
- STX10
- Gene Name
- syntaxin 10
There are no transcripts associated with this gene.
View Full Product Details