Product Details

SNP ID
rs2272320
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:101253126 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCGGGTCCGGGAGCGGAGGCCC[G/T]GGGGGCCCCGCGGGCAGGATGAGCT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CNOT11 PubMed Links

Gene Details

Gene
CNOT11
Gene Name
CCR4-NOT transcription complex subunit 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017546.4 244 Silent Mutation CCG,CCT P54P NP_060016.3

View Full Product Details