Product Details

SNP ID
rs1554777
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:101260947 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TAGTGAGCATTTCTCCTAGTAAGTG[A/G]ATGAGAAAAGATGCTATATGTCTAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNOT11 PubMed Links

Gene Details

Gene
CNOT11
Gene Name
CCR4-NOT transcription complex subunit 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017546.4 Intron NP_060016.3

View Full Product Details