Product Details

SNP ID
rs11961009
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:25081671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTGTAGAGAACTGTGGATGGAA[C/G]AAGTAAAATATTTTTGAATCTGGCT
Phenotype
MIM: 603209
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
CMAHP PubMed Links

Gene Details

Gene
CMAHP
Gene Name
cytidine monophospho-N-acetylneuraminic acid hydroxylase, pseudogene
There are no transcripts associated with this gene.

View Full Product Details