Product Details

SNP ID
rs11540586
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:29922811 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGAAATGCTTCTATATCCATCTATC[A/G]CCTCCTTGCCCTCTGTCTTCCCAAT
Phenotype
MIM: 614965 MIM: 612432
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SCRN1 PubMed Links

Gene Details

Gene
SCRN1
Gene Name
secernin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145513.1 2568 UTR 3 NP_001138985.1
NM_001145514.1 2568 UTR 3 NP_001138986.1
NM_001145515.1 2568 UTR 3 NP_001138987.1
NM_014766.4 2568 UTR 3 NP_055581.3
Gene
WIPF3
Gene Name
WAS/WASL interacting protein family member 3
There are no transcripts associated with this gene.

View Full Product Details