Product Details

SNP ID
rs2804916
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:13581612 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAAGGTATTAGTAGTTTGTTCTTA[C/T]TGCTGAAGAGTCTTCATGGTATGGA
Phenotype
MIM: 604483
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SIRT5 PubMed Links

Gene Details

Gene
SIRT5
Gene Name
sirtuin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193267.2 Intron NP_001180196.1
NM_001242827.1 Intron NP_001229756.1
NM_012241.4 Intron NP_036373.1
NM_031244.3 Intron NP_112534.1
XM_005248968.4 Intron XP_005249025.1
XM_011514444.2 Intron XP_011512746.1
XM_017010620.1 Intron XP_016866109.1
XM_017010621.1 Intron XP_016866110.1
XM_017010622.1 Intron XP_016866111.1
XM_017010623.1 Intron XP_016866112.1
XM_017010624.1 Intron XP_016866113.1
XM_017010625.1 Intron XP_016866114.1

View Full Product Details