Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001320800.1 | 2036 | Silent Mutation | CCC,CCT | P352P | NP_001307729.1 |
NM_007170.2 | 2036 | Silent Mutation | CCC,CCT | P435P | NP_009101.2 |
XM_005270355.3 | 2036 | Silent Mutation | CCC,CCT | P386P | XP_005270412.1 |
XM_006710287.2 | 2036 | Silent Mutation | CCC,CCT | P435P | XP_006710350.1 |
XM_011540497.1 | 2036 | Silent Mutation | CCC,CCT | P435P | XP_011538799.1 |
XM_011540500.2 | 2036 | Intron | XP_011538802.1 | ||
XM_017000047.1 | 2036 | Silent Mutation | CCC,CCT | P386P | XP_016855536.1 |
XM_017000048.1 | 2036 | Silent Mutation | CCC,CCT | P227P | XP_016855537.1 |
XM_017000049.1 | 2036 | Silent Mutation | CCC,CCT | P227P | XP_016855538.1 |