Product Details

SNP ID
rs1719134
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:36089600 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAAGTCATACCCCAACCCAAGAGAA[A/G]CCTTGGACATCTCTCATAAGACATC
Phenotype
MIM: 182283
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCL3 PubMed Links

Gene Details

Gene
CCL3
Gene Name
C-C motif chemokine ligand 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002983.2 Intron NP_002974.1

View Full Product Details