Product Details

SNP ID
rs139285
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.22:39099775 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GGGAGGGAGCACTCCCCTCCTCCTG[C/T]CCTCCTGCCAGCCCTTCCTTCCCCT
Phenotype
MIM: 610976
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APOBEC3H PubMed Links

Gene Details

Gene
APOBEC3H
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166002.2 Intron NP_001159474.2
NM_001166003.2 Intron NP_001159475.2
NM_001166004.2 Intron NP_001159476.2
NM_181773.4 Intron NP_861438.3
XM_011529990.2 Intron XP_011528292.1
XM_011529991.2 Intron XP_011528293.1
XM_011529992.2 Intron XP_011528294.1

View Full Product Details