Product Details

SNP ID
rs3915140
Assay Type
Functionally tested
NCBI dbSNP Submissions
74
Location
Chr.1:204033142 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCCAGGACAACGGTACAGCTGCTG[C/T]AGAGGCATAGGTTTGTCTGCTCTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00303 PubMed Links

Gene Details

Gene
LINC00303
Gene Name
long intergenic non-protein coding RNA 303
There are no transcripts associated with this gene.

View Full Product Details