Product Details

SNP ID
rs1009758
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:11139415 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TCCTCCGAGCCCCCTGCGTGTCGCT[C/G]TCCAGCTCTGCGGGGCTCCCCTGCC
Phenotype
MIM: 300118
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARHGAP6 PubMed Links

Gene Details

Gene
ARHGAP6
Gene Name
Rho GTPase activating protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287242.1 2023 Silent Mutation GAC,GAG D611E NP_001274171.1
NM_006125.2 2023 Intron NP_006116.2
NM_013423.2 2023 Silent Mutation GAC,GAG D588E NP_038267.1
NM_013427.2 2023 Silent Mutation GAC,GAG D791E NP_038286.2

View Full Product Details