Product Details
- SNP ID
-
rs808118
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:8538156 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCCAACCCAAGCCCAAGCCAAGGCC[A/G]CCATGTTTCTTTGGGAGTCTGAATT
- Phenotype
-
MIM: 300836
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANOS1
PubMed Links
Gene Details
- Gene
- ANOS1
- Gene Name
- anosmin 1
View Full Product Details