Product Details
- SNP ID
-
rs1180178
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:83193907 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- ATGCGGCCCAAGCGAAAAGGCCACC[C/G]CGGTGTTGGGGGGGGGCGGTCCGCG
- Phenotype
-
MIM: 172100
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PGM3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs16885957] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PGM3
- Gene Name
- phosphoglucomutase 3
- Gene
- RWDD2A
- Gene Name
- RWD domain containing 2A
View Full Product Details