Product Details
- SNP ID
-
rs1143832
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:64897995 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATGCAAATCTGAGTCTATCATTCCC[C/T]GTCTAAAAACTGTGTATGGCTTCCC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FLJ41278
PubMed Links
Gene Details
- Gene
- FLJ41278
- Gene Name
- uncharacterized LOC400046
There are no transcripts associated with this gene.
View Full Product Details