Product Details

SNP ID
rs938525
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:136863111 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTGAGGACGTAACCGTAGGAACGCT[C/T]TCGGTGTCAAACTGATGGACCCGGA
Phenotype
MIM: 600508
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NCK1 PubMed Links

Gene Details

Gene
NCK1
Gene Name
NCK adaptor protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190796.2 Intron NP_001177725.1
NM_001291999.1 Intron NP_001278928.1
NM_006153.5 Intron NP_006144.1
Gene
NCK1-AS1
Gene Name
NCK1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
SLC35G2
Gene Name
solute carrier family 35 member G2
There are no transcripts associated with this gene.

View Full Product Details