Product Details

SNP ID
rs1111375
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34521150 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATGGGTTCCAGTTTGCTTCCCCA[A/G]CCTAGAGGGCCTAGAGGTGCTCAGC
Phenotype
MIM: 604366
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAI1 PubMed Links

Gene Details

Gene
DNAI1
Gene Name
dynein axonemal intermediate chain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281428.1 970 Intron NP_001268357.1
NM_012144.3 970 Intron NP_036276.1
XM_006716758.3 970 Intron XP_006716821.1
XM_011517846.1 970 Intron XP_011516148.1
XM_011517847.2 970 Intron XP_011516149.1
XM_011517848.1 970 Intron XP_011516150.1
XM_011517849.1 970 Intron XP_011516151.1
XM_011517850.2 970 Intron XP_011516152.1
XM_017014625.1 970 Intron XP_016870114.1
Gene
ENHO
Gene Name
energy homeostasis associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198573.2 970 UTR 3 NP_940975.2

View Full Product Details