Product Details

SNP ID
rs1046002
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:110362953 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAAAGTAAGGATTACCTTTAATT[A/G]AGAACCTAAGTAATACAAAGAAAGT
Phenotype
MIM: 606077 MIM: 603878
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RBM15 PubMed Links

Gene Details

Gene
RBM15
Gene Name
RNA binding motif protein 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201545.1 3136 Intron NP_001188474.1
NM_022768.4 3136 Intron NP_073605.4
Gene
SLC16A4
Gene Name
solute carrier family 16 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201546.1 3136 UTR 3 NP_001188475.1
NM_001201547.1 3136 UTR 3 NP_001188476.1
NM_001201548.1 3136 UTR 3 NP_001188477.1
NM_001201549.1 3136 UTR 3 NP_001188478.1
NM_001319220.1 3136 UTR 3 NP_001306149.1
NM_004696.2 3136 UTR 3 NP_004687.1
XM_005271317.4 3136 Intron XP_005271374.1
XM_006711033.3 3136 UTR 3 XP_006711096.1

View Full Product Details