Product Details

SNP ID
rs1111179
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:41606749 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATCGGTTCAGAACCAGTCTGGCTG[G/T]GAGGCCCGAGGGCTGACAGCAGCGG
Phenotype
MIM: 612811
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LRFN5 PubMed Links

Gene Details

Gene
LRFN5
Gene Name
leucine rich repeat and fibronectin type III domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152447.3 67 Intron NP_689660.2
XM_005267369.3 67 UTR 5 XP_005267426.1
XM_017021048.1 67 Intron XP_016876537.1
XM_017021049.1 67 UTR 5 XP_016876538.1
XM_017021050.1 67 UTR 5 XP_016876539.1

View Full Product Details