Product Details

SNP ID
rs1063459
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:103278142 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGGCCATCCGGGCCAGCCGCGAG[C/G]AGATCCACGAGTATCGGCGCCAGCT
Phenotype
MIM: 605338
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
INA PubMed Links
Additional Information
For this assay, SNP(s) [rs7096855] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
INA
Gene Name
internexin neuronal intermediate filament protein alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032727.3 980 Missense Mutation CAG,GAG Q311E NP_116116.1

View Full Product Details