Product Details

SNP ID
rs882731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:94561507 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACAGACTGTGCCGTGAGTAACCCC[C/T]TGAGACAATGGAGACGAGAGACTCT
Phenotype
MIM: 147935
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SERPINA4 PubMed Links

Gene Details

Gene
SERPINA4
Gene Name
serpin family A member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289032.1 88 UTR 5 NP_001275961.1
NM_001289033.1 88 UTR 5 NP_001275962.1
NM_006215.3 88 Intron NP_006206.2

View Full Product Details