Product Details

SNP ID
hCV9599834
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:105925904 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCCAGCTTGCTCCTGGGTTCCAC[A/G]GCGGAGTCAGATGTTTCTGGGCAGT
Phenotype
MIM: 146910 MIM: 147070 MIM: 147010 MIM: 616623
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IGH PubMed Links

Gene Details

Gene
IGH
Gene Name
immunoglobulin heavy locus
There are no transcripts associated with this gene.

Gene
KIAA0125
Gene Name
KIAA0125
There are no transcripts associated with this gene.

View Full Product Details