Product Details

SNP ID
rs3910385
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:9577066 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGATACCAAAAATTTATTGTATGA[A/C]GTTTCAGATAAATTAATTCAGTTTA
Phenotype
MIM: 193525
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
WEE1 PubMed Links

Gene Details

Gene
WEE1
Gene Name
WEE1 G2 checkpoint kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143976.1 Intron NP_001137448.1
NM_003390.3 Intron NP_003381.1

View Full Product Details