Product Details

SNP ID
rs729369
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:8339202 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGGTTTGGGGAGCGATGGGGCTCC[A/G]AGGGATTTATTAATGCACGCCTCCG
Phenotype
MIM: 607670
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STK33 PubMed Links

Gene Details

Gene
STK33
Gene Name
serine/threonine kinase 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289058.1 Intron NP_001275987.1
NM_001289059.1 Intron NP_001275988.1
NM_001289061.1 Intron NP_001275990.1
NM_030906.3 Intron NP_112168.1
XM_011520288.2 Intron XP_011518590.1
XM_011520290.2 Intron XP_011518592.1
XM_011520292.2 Intron XP_011518594.1
XM_011520293.2 Intron XP_011518595.1
XM_011520294.1 Intron XP_011518596.1
XM_011520295.2 Intron XP_011518597.1
XM_011520297.2 Intron XP_011518599.1
XM_017018143.1 Intron XP_016873632.1
XM_017018144.1 Intron XP_016873633.1
XM_017018145.1 Intron XP_016873634.1
XM_017018146.1 Intron XP_016873635.1
XM_017018147.1 Intron XP_016873636.1
XM_017018148.1 Intron XP_016873637.1
XM_017018149.1 Intron XP_016873638.1
XM_017018150.1 Intron XP_016873639.1
XM_017018151.1 Intron XP_016873640.1
XM_017018152.1 Intron XP_016873641.1
XM_017018153.1 Intron XP_016873642.1
XM_017018154.1 Intron XP_016873643.1
XM_017018155.1 Intron XP_016873644.1
XM_017018156.1 Intron XP_016873645.1
XM_017018157.1 Intron XP_016873646.1
XM_017018158.1 Intron XP_016873647.1
XM_017018159.1 Intron XP_016873648.1
XM_017018160.1 Intron XP_016873649.1
XM_017018161.1 Intron XP_016873650.1
XM_017018162.1 Intron XP_016873651.1
XM_017018163.1 Intron XP_016873652.1
XM_017018164.1 Intron XP_016873653.1
XM_017018165.1 Intron XP_016873654.1

View Full Product Details