Product Details

SNP ID
rs1453416
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5803853 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATGTGCAGAGATAAAGAAAATGGA[A/G]CAAAATAACAATACAGGTGGGAGGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR52N2 PubMed Links

Gene Details

Gene
OR52N2
Gene Name
olfactory receptor family 52 subfamily N member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005174.1 421 Intron NP_001005174.1
XM_011520090.2 421 UTR 5 XP_011518392.1

View Full Product Details