Product Details

SNP ID
rs1048596
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6952106 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTACTACACTCAGCAGCAGGGTGT[C/G]CTGCAGGACCGCGACGGCACCATCA
Phenotype
MIM: 607462 MIM: 615140 MIM: 176883
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATN1 PubMed Links

Gene Details

Gene
ATN1
Gene Name
atrophin 1
There are no transcripts associated with this gene.

Gene
C12orf57
Gene Name
chromosome 12 open reading frame 57
There are no transcripts associated with this gene.

Gene
PTPN6
Gene Name
protein tyrosine phosphatase, non-receptor type 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002831.5 410 Silent Mutation GTC,GTG V85V NP_002822.2
NM_080548.4 410 Silent Mutation GTC,GTG V87V NP_536858.1
NM_080549.3 410 Silent Mutation GTC,GTG V85V NP_536859.1
XM_006718994.1 410 UTR 5 XP_006719057.1
XM_011520988.1 410 Silent Mutation GTC,GTG V87V XP_011519290.1
Gene
RNU7-1
Gene Name
RNA, U7 small nuclear 1
There are no transcripts associated with this gene.

View Full Product Details