Product Details
- SNP ID
-
rs1732287
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52378794 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTATTATGCCCCTTGGTGGTCTCCA[C/T]ATTTCCCCTAGCCCAGCACACAGGA
- Phenotype
-
MIM: 602766
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT84
PubMed Links
Gene Details
- Gene
- KRT84
- Gene Name
- keratin 84
View Full Product Details